Gene Transcripts

Gene Symbol
Bpifc
Entrez Gene ID
299685
Gene Name
BPI fold containing family C
Gene Aliases
Bpil2
Chromosome Location
Chr. 7: 23854846 - 23911953 on Build Rnor_6.0
UniGene ID
Rn.218679
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106773.1 NP_001100243.1 15 1455
XM_008765223.1 XP_008763445.1 14 1614
XM_008765224.1 XP_008763446.1 14 1608

View Full Product Details