Gene Transcripts

Gene Symbol
Ccdc53
Entrez Gene ID
299707
Gene Name
coiled-coil domain containing 53
Gene Aliases
RGD1563761
Chromosome Location
Chr. 7: 28762417 - 28809286 on Build Rnor_6.0
UniGene ID
Rn.8525
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106776.1 NP_001100246.1 4 403
XM_006241199.2 XP_006241261.1 4 375
GenBank mRNA FQ221863.1 - 3 178

View Full Product Details