Gene Transcripts

Gene Symbol
Satl1
Entrez Gene ID
302325
Gene Name
spermidine/spermine N1-acetyl transferase-like 1
Gene Aliases
RGD1565932
Chromosome Location
Chr. X: 83772526 - 83788430 on Build Rnor_6.0
UniGene ID
Rn.127182
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163543.1 NP_001157015.1 1 1193
XM_006257190.2 XP_006257252.1 1 1640

View Full Product Details