Gene Transcripts

Gene Symbol
Slc9a6
Entrez Gene ID
302863
Gene Name
solute carrier family 9, subfamily A (NHE6, cation proton antiporter 6), member 6
Gene Aliases
RGD1563582
Chromosome Location
Chr. X: 158988840 - 159045017 on Build Rnor_6.0
UniGene ID
Rn.25009
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773665.1 XP_008771887.1 13 1490
XM_217630.8 XP_217630.3 12 1530

View Full Product Details