Gene Transcripts

Gene Symbol
Hs3st3b1
Entrez Gene ID
303218
Gene Name
heparan sulfate (glucosamine) 3-O-sulfotransferase 3B1
Gene Aliases
Hs3st3b
Chromosome Location
Chr. 10: 50369097 - 50402584 on Build Rnor_6.0
UniGene ID
Rn.231429
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191646.1 NP_001178575.1 1 579

View Full Product Details