Gene Transcripts

Gene Symbol
Lmod1
Entrez Gene ID
304816
Gene Name
leiomodin 1 (smooth muscle)
Gene Aliases
Chromosome Location
Chr. 13: 52147140 - 52189689 on Build Rnor_6.0
UniGene ID
Rn.66554
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107179.1 NP_001100649.1 2 1224
XM_008769562.1 XP_008767784.1 2 1652

View Full Product Details