Gene Transcripts

Gene Symbol
Adnp2
Entrez Gene ID
307236
Gene Name
ADNP homeobox 2
Gene Aliases
RGD1308279
Chromosome Location
Chr. 18: 76637415 - 76661186 on Build Rnor_6.0
UniGene ID
Rn.17024
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127373.1 NP_001120845.1 4 1001
GenBank mRNA BC166606.1 - 4 1001

View Full Product Details