Gene Transcripts

Gene Symbol
Ogfod1
Entrez Gene ID
307657
Gene Name
2-oxoglutarate and iron-dependent oxygenase domain containing 1
Gene Aliases
RGD1308848
Chromosome Location
Chr. 19: 11425301 - 11451085 on Build Rnor_6.0
UniGene ID
Rn.228470
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107411.2 NP_001100881.1 7 675

View Full Product Details