Gene Transcripts

Gene Symbol
Zbtb45
Entrez Gene ID
308366
Gene Name
zinc finger and BTB domain containing 45
Gene Aliases
Zfp499, Znf499
Chromosome Location
Chr. 1: 65562874 - 65568301 on Build Rnor_6.0
UniGene ID
Rn.203449
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107478.1 NP_001100948.1 2 943
XM_006228114.2 XP_006228176.1 2 2204
XM_006228115.2 XP_006228177.1 2 2112

View Full Product Details