Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014019.1 | NP_001014041.1 | 4 | 1041 | |
XM_006257219.2 | XP_006257281.1 | 4 | 1084 | ||
XM_006257220.2 | XP_006257282.1 | 3 | 1056 | ||
XM_006257221.2 | XP_006257283.1 | 3 | 1040 | ||
GenBank mRNA | BC079240.1 | - | 4 | 1041 |