Gene Transcripts

Gene Symbol
Art1
Entrez Gene ID
308873
Gene Name
ADP-ribosyltransferase 1
Gene Aliases
Chromosome Location
Chr. 1: 167202064 - 167206615 on Build Rnor_6.0
UniGene ID
Rn.6493
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107541.2 NP_001101011.1 2 859

View Full Product Details