Gene Transcripts

Gene Symbol
Scgb1c1
Entrez Gene ID
309100
Gene Name
secretoglobin, family 1C, member 1
Gene Aliases
RGD1561432, Ryd5
Chromosome Location
Chr. 1: 213595240 - 213596459 on Build Rnor_6.0
UniGene ID
Rn.41707
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107561.1 NP_001101031.1 2 252
GenBank mRNA X60661.1 - 2 231

View Full Product Details