Gene Transcripts

Gene Symbol
Gopc
Entrez Gene ID
309774
Gene Name
golgi associated PDZ and coiled-coil motif containing
Gene Aliases
Chromosome Location
Chr. 20: 33471355 - 33521281 on Build Rnor_6.0
UniGene ID
Rn.27865
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107631.1 NP_001101101.1 8 1354
GenBank mRNA BC168160.1 - 8 1383

View Full Product Details