Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003587.4 | NP_003578.2 | 1, 2 | 420 | |
GenBank mRNA | AB001601.1 | - | 1, 2 | 347 | |
AK312264.1 | - | 1, 2 | 359 | ||
BC002789.2 | - | 1, 2 | 343 | ||
BC008825.2 | - | 1, 2 | 371 | 4100320 | |
BC009392.2 | - | 1, 2 | 371 | 4136069 | |
DB072495.1 | - | 1, 2 | 421 |