Gene Transcripts

Gene Symbol
Otud7b
Entrez Gene ID
310677
Gene Name
OTU deubiquitinase 7B
Gene Aliases
RGD1308111
Chromosome Location
Chr. 2: 198231353 - 198300201 on Build Rnor_6.0
UniGene ID
Rn.199225
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107697.1 NP_001101167.1 8 1228
XM_006232957.2 XP_006233019.1 8 997
XM_006232958.2 XP_006233020.1 7 805

View Full Product Details