Gene Transcripts

Gene Symbol
Prcc
Entrez Gene ID
310687
Gene Name
papillary renal cell carcinoma (translocation-associated)
Gene Aliases
Chromosome Location
Chr. 2: 187232458 - 187258140 on Build Rnor_6.0
UniGene ID
Rn.12346
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107700.1 NP_001101170.1 6 1467
GenBank mRNA BC166708.1 - 6 1467

View Full Product Details