Gene Transcripts

Gene Symbol
Rsbn1
Entrez Gene ID
310749
Gene Name
round spermatid basic protein 1
Gene Aliases
RGD1307231
Chromosome Location
Chr. 2: 206392200 - 206451671 on Build Rnor_6.0
UniGene ID
Rn.38954
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191710.1 NP_001178639.1 7 2500
XM_006233082.2 XP_006233144.1 7 2199

View Full Product Details