Gene Transcripts

Gene Symbol
Stk35
Entrez Gene ID
311419
Gene Name
serine/threonine kinase 35
Gene Aliases
Stk35l1
Chromosome Location
Chr. 3: 122307967 - 122340205 on Build Rnor_6.0
UniGene ID
Rn.18285
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107773.1 NP_001101243.1 3 2483
XM_006234974.2 XP_006235036.1 3 1431

View Full Product Details