Gene Transcripts
- Gene Symbol
- Doxl1
- Entrez Gene ID
- 312316
- Gene Name
- diamine oxidase-like protein 1
- Gene Aliases
- Chromosome Location
- Chr. 4: 78563625 - 78567241 on Build Rnor_6.0
- UniGene ID
- Rn.79162
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_199233.1 |
NP_954703.1
|
1 |
226 |
|
View Full Product Details