Gene Transcripts

Gene Symbol
Spocd1
Entrez Gene ID
313060
Gene Name
SPOC domain containing 1
Gene Aliases
RGD1560989, Spocd1-ps1
Chromosome Location
Chr. 5: 148145416 - 148166567 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764193.1 XP_008762415.1 12 1564

View Full Product Details