Gene Transcripts
- Gene Symbol
- Dfnb31
- Entrez Gene ID
- 313255
- Gene Name
- deafness, autosomal recessive 31
- Gene Aliases
- Cip98, Whrn
- Chromosome Location
- Chr. 5: 79235541 - 79317206 on Build Rnor_6.0
- UniGene ID
- Rn.231801
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181088.1 |
NP_851602.1
|
12 |
2569 |
|
GenBank mRNA |
AY227205.1 |
- |
12 |
2569 |
|
View Full Product Details