Gene Transcripts

Gene Symbol
Dock11
Entrez Gene ID
313438
Gene Name
dedicator of cytokinesis 11
Gene Aliases
Chromosome Location
Chr. X: 122507171 - 122690463 on Build Rnor_6.0
UniGene ID
Rn.22563
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191760.1 NP_001178689.1 21 2455
XM_006257453.2 XP_006257515.1 21 2658

View Full Product Details