Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136540.1 | NP_001130012.1 | 6 | 652 | |
NM_001136541.1 | NP_001130013.1 | 5 | 559 | ||
NM_003661.3 | NP_003652.2 | 6 | 613 | ||
NM_145343.2 | NP_663318.1 | 7 | 767 | ||
XM_005261796.3 | XP_005261853.1 | 5 | 538 | ||
XM_011530478.2 | XP_011528780.1 | 6 | 685 | ||
GenBank mRNA | AF305224.1 | - | 6 | 552 | |
AF305428.1 | - | 6 | 592 | ||
AK296099.1 | - | 7 | 750 | ||
BC017331.1 | - | 6 | 543 | ||
BC143039.1 | - | 6 | 543 | 40034506 | |
BG419709.1 | - | 5 | 482 | ||
KU178424.1 | - | 5 | 445 | ||
KU178425.1 | - | 4 | 391 |