Gene Transcripts

Gene Symbol
Klhl15
Entrez Gene ID
314111
Gene Name
kelch-like family member 15
Gene Aliases
RGD1563101
Chromosome Location
Chr. X: 63343006 - 63394190 on Build Rnor_6.0
UniGene ID
Rn.100658
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108021.1 NP_001101491.1 3 1296
XM_006257022.2 XP_006257084.1 6 1571
XM_006257023.2 XP_006257085.1 3 1696
XM_006257024.2 XP_006257086.1 4 1747
XM_006257027.2 XP_006257089.1 3 2625
XM_008773257.1 XP_008771479.1 2 3416
XM_008773259.1 XP_008771481.1 5 1791
XM_008773260.1 XP_008771482.1 2 3416

Gene Symbol
LOC100911168
Entrez Gene ID
100911168
Gene Name
kelch-like protein 15-like
Gene Aliases
Chromosome Location
Chr. X: 63392209 - 63400743 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257037.1 XP_006257099.1 2 527

View Full Product Details