Gene Transcripts

Gene Symbol
Ak7
Entrez Gene ID
314416
Gene Name
adenylate kinase 7
Gene Aliases
Chromosome Location
Chr. 6: 129538645 - 129606939 on Build Rnor_6.0
UniGene ID
Rn.162876
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108055.3 NP_001101525.2 11 1321
XM_008764805.1 XP_008763027.1 11 1751
XM_008764806.1 XP_008763028.1 10 1670

View Full Product Details