Gene Transcripts

Gene Symbol
Dennd3
Entrez Gene ID
315055
Gene Name
DENN/MADD domain containing 3
Gene Aliases
RGD1311559
Chromosome Location
Chr. 7: 114724554 - 114812471 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006241738.1 XP_006241800.1 17 3223
XM_008765603.1 XP_008763825.1 17 3262
XM_235398.6 XP_235398.6 17 3262

View Full Product Details