Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_006241927.2 | XP_006241989.1 | 3, 4 | 446 | |
XM_008765571.1 | XP_008763793.1 | 3, 4 | 443 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_003750369.3 | XP_003750417.2 | 3, 4 | 446 | |
XM_008765633.1 | XP_008763855.1 | 3, 4 | 443 |