Gene Transcripts

Gene Symbol
Ky
Entrez Gene ID
315962
Gene Name
kyphoscoliosis peptidase
Gene Aliases
Chromosome Location
Chr. 8: 110982777 - 111022666 on Build Rnor_6.0
UniGene ID
Rn.24084
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108180.1 NP_001101650.1 4, 5 740
XM_006243657.2 XP_006243719.1 4, 5 740

View Full Product Details