Gene Transcripts

Gene Symbol
ALDH4A1
Entrez Gene ID
8659
Gene Name
aldehyde dehydrogenase 4 family member A1
Gene Aliases
ALDH4, P5CD, P5CDh
Chromosome Location
Chr. 1: 18871430 - 18902799 on Build GRCh38
UniGene ID
Hs.77448
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161504.1 NP_001154976.1 14 1652
NM_001319218.1 NP_001306147.1 13 1639
NM_003748.3 NP_003739.2 14 1792
NM_170726.2 NP_733844.1 14 1792
GenBank mRNA AK222486.1 - 14 1548
AK289972.1 - 14 1718
AK294552.1 - 14 1652
BC007581.1 - 14 1522 3139944
BC023600.2 - 14 1533 4548787
BX649081.1 - 1 897
FJ462711.1 - 13 1377
U24266.1 - 14 1546
U24267.1 - 14 1546

View Full Product Details