Gene Transcripts

Gene Symbol
IRS2
Entrez Gene ID
8660
Gene Name
insulin receptor substrate 2
Gene Aliases
IRS-2
Chromosome Location
Chr. 13: 109753837 - 109786567 on Build GRCh38
UniGene ID
Hs.442344
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003749.2 NP_003740.2 2 4981
GenBank mRNA AF073310.1 - 2 4981
BC026044.1 - 2 497
BC066356.1 - 2 476
CA952668.1 - 1 484

View Full Product Details