Gene Transcripts

Gene Symbol
Ube2q2l
Entrez Gene ID
317341
Gene Name
ubiquitin-conjugating enzyme E2Q family member 2-like
Gene Aliases
RGD1563226
Chromosome Location
Chr. X: 10771112 - 10772633 on Build Rnor_6.0
UniGene ID
Rn.106891
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135997.2 NP_001129469.1 1 682
GenBank mRNA FQ225248.1 - 1 129
FQ225615.1 - 1 129

View Full Product Details