Gene Transcripts
- Gene Symbol
- Lhfpl4
- Entrez Gene ID
- 353230
- Gene Name
- lipoma HMGIC fusion partner-like 4
- Gene Aliases
- Chromosome Location
- Chr. 4: 145117533 - 145147397 on Build Rnor_6.0
- UniGene ID
- Rn.145133
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181387.2 |
NP_852052.1
|
3, 4 |
832 |
|
GenBank mRNA |
AY278321.1 |
- |
3, 4 |
832 |
|
View Full Product Details