Gene Transcripts

Gene Symbol
Trim37
Entrez Gene ID
360592
Gene Name
tripartite motif-containing 37
Gene Aliases
Chromosome Location
Chr. 10: 74436129 - 74568636 on Build Rnor_6.0
UniGene ID
Rn.228843
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108288.1 NP_001101758.1 17 1961
XM_006247099.2 XP_006247161.1 16 2047
XM_006247100.2 XP_006247162.1 16 2046
XM_006247101.2 XP_006247163.1 16 2047
XM_006247102.2 XP_006247164.1 16 2046
XM_006247104.2 XP_006247166.1 16 2046
XM_006247105.2 XP_006247167.1 16 2046
XM_008768074.1 XP_008766296.1 16 2046
GenBank mRNA BC168955.1 - 16 1882

View Full Product Details