Gene Transcripts

Gene Symbol
Ten1
Entrez Gene ID
360664
Gene Name
TEN1 CST complex subunit
Gene Aliases
RGD1311078
Chromosome Location
Chr. 10: 105073077 - 105095094 on Build Rnor_6.0
UniGene ID
Rn.15123
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108307.1 NP_001101777.1 3 295

View Full Product Details