Gene Transcripts

Gene Symbol
SNX3
Entrez Gene ID
8724
Gene Name
sorting nexin 3
Gene Aliases
Grd19, MCOPS8, SDP3
Chromosome Location
Chr. 6: 108211217 - 108261260 on Build GRCh38
UniGene ID
Hs.12102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300928.1 NP_001287857.1 2 575
NM_001300929.1 NP_001287858.1 2 509
NM_003795.5 NP_003786.1 2 575
GenBank mRNA AF034546.1 - 2 279
AF062483.1 - 2 255
AK289476.1 - 2 366
BC008444.1 - 2 333 4102735
BC014580.1 - 2 357 3932208
BC015179.1 - 2 339 3905515
BC016863.2 - 2 589 3846511
BT007114.1 - 2 236
CN422502.1 - 2 255
CR456898.1 - 2 236
HY156437.1 - 2 413

View Full Product Details