Gene Transcripts

Gene Symbol
Spetex-2G
Entrez Gene ID
361099
Gene Name
Spetex-2G protein
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.202552
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009971.1 NP_001009971.1 4 541
GenBank mRNA AB180082.1 - 4 541

Gene Symbol
Spetex-2A
Entrez Gene ID
364261
Gene Name
Spetex-2A protein
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.198259
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009976.1 NP_001009976.1 4 586
GenBank mRNA AB180076.1 - 4 586

Gene Symbol
Spetex-2B
Entrez Gene ID
497084
Gene Name
Spetex-2B protein
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.198259
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011697.1 NP_001011697.1 4 541
GenBank mRNA AB180077.1 - 4 541

Gene Symbol
Spetex-2C
Entrez Gene ID
364255
Gene Name
Spetex-2C protein
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.215809
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011698.1 NP_001011698.1 4 541
GenBank mRNA AB180078.1 - 4 541

Gene Symbol
Spetex-2E
Entrez Gene ID
361012
Gene Name
Spetex-2E protein
Gene Aliases
Chromosome Location
Chr. 15: 5414416 - 5417642 on Build Rnor_6.0
UniGene ID
Rn.129858
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011702.1 NP_001011702.1 541
GenBank mRNA AB180080.1 - 541

Gene Symbol
Spetex-2F
Entrez Gene ID
316940
Gene Name
Spetex-2F protein
Gene Aliases
Chromosome Location
Chr. 15: 5463852 - 5509839 on Build Rnor_6.0
UniGene ID
Rn.110794
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009968.1 NP_001009968.1 4 541
GenBank mRNA AB180081.1 - 4 541

Gene Symbol
Spetex-2H
Entrez Gene ID
361008
Gene Name
Spetex-2H protein
Gene Aliases
Spergen-2H
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.162039
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009969.1 NP_001009969.1 4 547
GenBank mRNA AB180083.1 - 4 547

View Full Product Details