Gene Transcripts

Gene Symbol
Nudt5
Entrez Gene ID
361274
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 5
Gene Aliases
Chromosome Location
Chr. 17: 76387027 - 76410313 on Build Rnor_6.0
UniGene ID
Rn.55388
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007733.1 NP_001007734.1 3 234
XM_008771875.1 XP_008770097.1 3 253
GenBank mRNA BC079176.1 - 3 234

View Full Product Details