Gene Transcripts

Gene Symbol
Rps6kb2
Entrez Gene ID
361696
Gene Name
ribosomal protein S6 kinase, polypeptide 2
Gene Aliases
Chromosome Location
Chr. 1: 219406201 - 219412888 on Build Rnor_6.0
UniGene ID
Rn.162512
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010962.2 NP_001010962.2 10 910
GenBank mRNA BC089102.1 - 10 915
DY561917.1 - 4 285

View Full Product Details