Gene Transcripts

Gene Symbol
Zdhhc5
Entrez Gene ID
362156
Gene Name
zinc finger, DHHC-type containing 5
Gene Aliases
DHHC-5
Chromosome Location
Chr. 3: 72091314 - 72113680 on Build Rnor_6.0
UniGene ID
Rn.234348
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039338.1 NP_001034427.1 10 1852
GenBank mRNA AY871203.1 - 10 1852

View Full Product Details