Gene Transcripts

Gene Symbol
Loh12cr1
Entrez Gene ID
362452
Gene Name
loss of heterozygosity, 12, chromosomal region 1
Gene Aliases
Chromosome Location
Chr. 4: 168396042 - 168466917 on Build Rnor_6.0
UniGene ID
Rn.53228
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108650.2 NP_001102120.1 2 162
XM_006237528.2 XP_006237590.1 2 895
XM_008763394.1 XP_008761616.1 2 893
XM_008763395.1 XP_008761617.1 2 893
XM_008763396.1 XP_008761618.1 2 895
GenBank mRNA BC160872.1 - 2 162
FQ211769.1 - 2 257

View Full Product Details