Gene Transcripts

Gene Symbol
Fkbp15
Entrez Gene ID
362528
Gene Name
FK506 binding protein 15
Gene Aliases
RGD1304816
Chromosome Location
Chr. 5: 78133357 - 78229311 on Build Rnor_6.0
UniGene ID
Rn.162820
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008763840.1 XP_008762062.1 7, 8 740
GenBank mRNA FQ230831.1 - 6, 7 667

View Full Product Details