Gene Transcripts

Gene Symbol
Alg6
Entrez Gene ID
362547
Gene Name
ALG6, alpha-1,3-glucosyltransferase
Gene Aliases
Chromosome Location
Chr. 5: 118415680 - 118470634 on Build Rnor_6.0
UniGene ID
Rn.104225
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033709.1 NP_001028881.1 1007
GenBank mRNA BC101850.1 - 1007
FQ210318.1 - 1022

View Full Product Details