Gene Transcripts

Gene Symbol
SPHK1
Entrez Gene ID
8877
Gene Name
sphingosine kinase 1
Gene Aliases
SPHK
Chromosome Location
Chr. 17: 76382977 - 76387860 on Build GRCh38
UniGene ID
Hs.68061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142601.1 NP_001136073.1 6 1469
NM_001142602.1 NP_001136074.1 5 1514
NM_021972.3 NP_068807.2 6 1511
NM_182965.2 NP_892010.2 6 1832
XM_005257766.2 XP_005257823.1 6 1824
GenBank mRNA AF200328.1 - 5 1095
AF238083.1 - 6 1413
AF266756.1 - 5 1346
AJ245504.1 - 4 1068
AK022402.1 - 6 1469
AK023393.1 - 5 1426
AK095578.1 - 5 2154
AK122781.1 - 8 1886
AK225250.1 - 6 1432
AK225258.1 - 6 1298
AK292294.1 - 6 1556
AK292614.1 - 5 1430
BC004112.2 - 2 786
BC008040.2 - 6 1406 3831657
BC009419.1 - 6 1499 3506924
BC014439.1 - 6 1769 4871343
BC030553.1 - 6 1791 5213270

View Full Product Details