Gene Transcripts

Gene Symbol
Ttc7a
Entrez Gene ID
362696
Gene Name
tetratricopeptide repeat domain 7A
Gene Aliases
Ttc7
Chromosome Location
Chr. 6: 10912383 - 11014279 on Build Rnor_6.0
UniGene ID
Rn.16768
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100756.1 NP_001094226.1 5 946
GenBank mRNA BC168226.1 - 5 946

View Full Product Details