Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317086.1 | NP_001304015.1 | 1 | 202 | |
NM_003904.4 | NP_003895.1 | 1 | 202 | ||
GenBank mRNA | AF019767.1 | - | 1 | 182 | |
AK301227.1 | - | 1 | 150 | ||
BC004256.1 | - | 1 | 157 | 3610932 | |
BC012162.1 | - | 1 | 170 | 4578136 | |
BC017349.2 | - | 1 | 187 | 5013820 | |
BC017380.2 | - | 1 | 170 | 2988688 | |
BC111028.1 | - | 1 | 174 | 5245870 | |
BT006642.1 | - | 1 | 154 | ||
CD359945.1 | - | 1 | 202 | ||
CR975994.1 | - | 1 | 34 |