Gene Transcripts

Gene Symbol
Eml1
Entrez Gene ID
362783
Gene Name
echinoderm microtubule associated protein like 1
Gene Aliases
Chromosome Location
Chr. 6: 132367342 - 132450488 on Build Rnor_6.0
UniGene ID
Rn.211693
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025741.1 NP_001020912.1 3 413
GenBank mRNA BC097450.1 - 3 413

View Full Product Details