Gene Transcripts
- Gene Symbol
- Gnptab
- Entrez Gene ID
- 362865
- Gene Name
- N-acetylglucosamine-1-phosphate transferase, alpha and beta subunits
- Gene Aliases
- RGD1564821
- Chromosome Location
- Chr. 7: 28956351 - 29022183 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_343194.7 |
XP_343195.3
|
19 |
3709 |
|
View Full Product Details