Gene Transcripts

Gene Symbol
Emc2
Entrez Gene ID
362905
Gene Name
ER membrane protein complex subunit 2
Gene Aliases
RGD1310430, Ttc35
Chromosome Location
Chr. 7: 82338639 - 82377389 on Build Rnor_6.0
UniGene ID
Rn.203197
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113785.1 NP_001107257.1 8, 9 723
GenBank mRNA BC158805.1 - 8, 9 723
FQ214096.1 - 5, 6 606
FQ215024.1 - 8, 9 712

View Full Product Details