Gene Transcripts

Gene Symbol
Nudcd1
Entrez Gene ID
362906
Gene Name
NudC domain containing 1
Gene Aliases
RGD1310624
Chromosome Location
Chr. 7: 83302778 - 83348529 on Build Rnor_6.0
UniGene ID
Rn.228609
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130561.2 NP_001124033.1 5 796
XM_006241614.2 XP_006241676.1 5 895
GenBank mRNA BC169014.1 - 5 796

View Full Product Details