Gene Transcripts

Gene Symbol
Rsl24d1
Entrez Gene ID
363099
Gene Name
ribosomal L24 domain containing 1
Gene Aliases
RGD1309784
Chromosome Location
Chr. 8: 79792587 - 79801592 on Build Rnor_6.0
UniGene ID
Rn.105523
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014212.1 NP_001014234.1 1 116
GenBank mRNA BC062237.1 - 1 116

View Full Product Details